missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF71 Full-length ORF (NP_067039.1, 1 a.a. - 489 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00058491-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MKELDPKNDISEDKLSVVGEATGGPTRNGARGPGSEGVWEPGSWPERPRGDAGAEWEPLGIPQGNKLLGGSVPACHELKAFANQGCVLVPPRLDDPTEKGACPPVRRGKNFSSTSDLSKPPMPCEEKKTYDCSECGKAFSRSSSLIKHQRIHTGEKPFECDTCGKHFIERSSLTIHQRVHTGEKPYACGDCGKAFSQRMNLTVHQRTHTGEKPYVCDVCGKAFRKTSSLTQHERIHTGEKPYACGDCGKAFSQNMHLIVHQRTHTGEKPYVCPECGRAFSQNMHLTEHQRTHTGEKPYACKECGKAFNKSSSLTLHQRNHTGEKPYVCGECGKAFSQSSYLIQHQRFHIGVKPFECSECGKAFSKNSSLTQHQRIHTGEKPYECYICKKHFTGRSSLIVHQIVHTGEKPYVCGECGKAFSQSAYLIEHQRIHTGEKPYRCGQCGKSFIKNSSLTVHQRIHTGEKPYRCGECGKTFSRNTNLTRHLRIHTSpecifications
NP_067039.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EZFIT | |
ZNF71 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
58491 | |
ZNF71 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKELDPKNDISEDKLSVVGEATGGPTRNGARGPGSEGVWEPGSWPERPRGDAGAEWEPLGIPQGNKLLGGSVPACHELKAFANQGCVLVPPRLDDPTEKGACPPVRRGKNFSSTSDLSKPPMPCEEKKTYDCSECGKAFSRSSSLIKHQRIHTGEKPFECDTCGKHFIERSSLTIHQRVHTGEKPYACGDCGKAFSQRMNLTVHQRTHTGEKPYVCDVCGKAFRKTSSLTQHERIHTGEKPYACGDCGKAFSQNMHLIVHQRTHTGEKPYVCPECGRAFSQNMHLTEHQRTHTGEKPYACKECGKAFNKSSSLTLHQRNHTGEKPYVCGECGKAFSQSSYLIQHQRFHIGVKPFECSECGKAFSKNSSLTQHQRIHTGEKPYECYICKKHFTGRSSLIVHQIVHTGEKPYVCGECGKAFSQSAYLIEHQRIHTGEKPYRCGQCGKSFIKNSSLTVHQRIHTGEKPYRCGECGKTFSRNTNLTRHLRIHT | |
RUO | |
ZNF71 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |