missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF690 Partial ORF (NP_689668, 101 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00146050-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192027

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

Sequence: RPRSSVTVSVKGQEVRLEKMTPPKSSQELLSVRQESVEPQPRGVPKKERARSPDLGPQEQMNPKEKLKPFQRSGLPFPKSGVVSRLEQGEPWIPDLLGS
Specifications

Specifications

NP_689668
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RPRSSVTVSVKGQEVRLEKMTPPKSSQELLSVRQESVEPQPRGVPKKERARSPDLGPQEQMNPKEKLKPFQRSGLPFPKSGVVSRLEQGEPWIPDLLGS
RUO
ZSCAN29
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
146050
ZNF690 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ35867/MGC129894/MGC129895/ZNF690/Zfp690
ZSCAN29
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers