missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF644 Full-length ORF (NP_057704.2, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084146-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195653

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MLIRQNLALDCKQKKSRSRSGSKKKMLTLPHGADEVYILRCRFCGLVFRGPLSVQEDWIKHLQRHIVNANLPRTGAGMVEVTSLLKKPASITETSFSLLMAEAAS
Specifications

Specifications

NP_057704.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BM-005/KIAA1221/MGC60165/MGC70410/Zep-2
ZNF644
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84146
ZNF644 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLIRQNLALDCKQKKSRSRSGSKKKMLTLPHGADEVYILRCRFCGLVFRGPLSVQEDWIKHLQRHIVNANLPRTGAGMVEVTSLLKKPASITETSFSLLMAEAAS
RUO
ZNF644
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers