missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF484 Full-length ORF (NP_113674.1, 1 a.a. - 852 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083744-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16105473

  • 349.00€ / 10µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

Sequence: MTKSLESVSFKDVTVDFSRDEWQQLDLAQKSLYREVMLENYFNLISVGCQVPKPEVIFSLEQEEPCMLDGEIPSQSRPDGDIGFGPLQQRMSEEVSFQSEININLFTRDDPYSILEELWKDDEHTRKCGENQNKPLSRVVFINKKTLANDSIFEYKDIGEIVHVNTHLVSSRKRPHNCNSCGKNLEPIITLYNRNNATENSDKTIGDGDIFTHLNSHTEVTACECNQCGKPLHHKQALIQQQKIHTRESLYLFSDYVNVFSPKSHAFAHESICAEEKQHECHECEAVFTQKSQLDGSQRVYAGICTEYEKDFSLKSNRQKTPYEGNYYKCSDYGRAFIQKSDLFRCQRIHSGEKPYEYSECEKNLPQNSNLNIHKKIHTGGKHFECTECGKAFTRKSTLSMHQKIHTGEKPYVCTECGKAFIRKSHFITHERIHTGEKPYECSDCGKSFIKKSQLHVHQRIHTGENPFICSECGKVFTHKTNLIIHQKIHTGERPYICTVCGKAFTDRSNLIKHQKIHTGEKPYKCSDCGKSFTWKSRLRIHQKCHTGERHYECSECGKAFIQKSTLSMHQRIHRGEKPYVCTECGKAFFHKSHFITHERIHTGEKPYECSICGKSFTKKSQLHVHQQIHTGEKPYRCAECGKAFTDRSNLFTHQKIHTGEKPYKCSDCGKAFTRKSGLHIHQQSHTGERHYECSECGKAFARKSTLIMHQRIHTGEKPYICNECGKSFIQKSHLNRHRRIHTGEKPYECSDCGKSFIKKSQLHEHHRIHTGEKPYICAECGKAFTIRSNLIKHQKIHTKQKPYKCSDLGKALNWKPQLSMPQKSDNGEVECSMPQLWCGDSEGDQGQLSSI
Specifications

Specifications

NP_113674.1
Liquid
83744
ZNF484 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTKSLESVSFKDVTVDFSRDEWQQLDLAQKSLYREVMLENYFNLISVGCQVPKPEVIFSLEQEEPCMLDGEIPSQSRPDGDIGFGPLQQRMSEEVSFQSEININLFTRDDPYSILEELWKDDEHTRKCGENQNKPLSRVVFINKKTLANDSIFEYKDIGEIVHVNTHLVSSRKRPHNCNSCGKNLEPIITLYNRNNATENSDKTIGDGDIFTHLNSHTEVTACECNQCGKPLHHKQALIQQQKIHTRESLYLFSDYVNVFSPKSHAFAHESICAEEKQHECHECEAVFTQKSQLDGSQRVYAGICTEYEKDFSLKSNRQKTPYEGNYYKCSDYGRAFIQKSDLFRCQRIHSGEKPYEYSECEKNLPQNSNLNIHKKIHTGGKHFECTECGKAFTRKSTLSMHQKIHTGEKPYVCTECGKAFIRKSHFITHERIHTGEKPYECSDCGKSFIKKSQLHVHQRIHTGENPFICSECGKVFTHKTNLIIHQKIHTGERPYICTVCGKAFTDRSNLIKHQKIHTGEKPYKCSDCGKSFTWKSRLRIHQKCHTGERHYECSECGKAFIQKSTLSMHQRIHRGEKPYVCTECGKAFFHKSHFITHERIHTGEKPYECSICGKSFTKKSQLHVHQQIHTGEKPYRCAECGKAFTDRSNLFTHQKIHTGEKPYKCSDCGKAFTRKSGLHIHQQSHTGERHYECSECGKAFARKSTLIMHQRIHTGEKPYICNECGKSFIQKSHLNRHRRIHTGEKPYECSDCGKSFIKKSQLHEHHRIHTGEKPYICAECGKAFTIRSNLIKHQKIHTKQKPYKCSDLGKALNWKPQLSMPQKSDNGEVECSMPQLWCGDSEGDQGQLSSI
RUO
ZNF484
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
124.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BA526D8.4/MGC133033/MGC133034
ZNF484
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers