missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF483 Partial ORF (NP_001007170.1, 142 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00158399-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192217

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Sequence: QDSTVSQEENSKEDKMVTVCPNTESCESITLKDVAVNFSRGEWKKLEPFQKELYKEVLLENLRNLEFLDFPVSKLELISQLKWVELPWL
Specifications

Specifications

NP_001007170.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QDSTVSQEENSKEDKMVTVCPNTESCESITLKDVAVNFSRGEWKKLEPFQKELYKEVLLENLRNLEFLDFPVSKLELISQLKWVELPWL
RUO
ZNF483
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
158399
ZNF483 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ZKSCAN16
ZNF483
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers