missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF446 Partial ORF (NP_060378, 351 a.a. - 449 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055663-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: HRTHTSGPGVQSPGLATGESTEKPPQGEVAFPHHPRRSLTGPRSYPCEECGCSFSWKSQLVIHRKSHTGQRRHFCSDCGRAFDWKSQLVIHRKGHRPEVSpecifications
NP_060378 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HRTHTSGPGVQSPGLATGESTEKPPQGEVAFPHHPRRSLTGPRSYPCEECGCSFSWKSQLVIHRKSHTGQRRHFCSDCGRAFDWKSQLVIHRKGHRPEV | |
RUO | |
ZNF446 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55663 | |
ZNF446 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20626/ZKSCAN20/ZSCAN30 | |
ZNF446 | |
Recombinant | |
wheat germ expression system |