missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF32 Partial ORF (NP_001005368.1, 173 a.a. - 273 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007580-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16127725

  • 335.00€ / 10µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Sequence: QSNLAVHRRVHSGEKPYRCDQCGKAFSQKGSLIVHIRVHTGLKPYACTQCRKSFHTRGNCILHGKIHTGETPYLCGQCGKSFTQRGSLAVHQRSCSQRLTL
Specifications

Specifications

NP_001005368.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QSNLAVHRRVHSGEKPYRCDQCGKAFSQKGSLIVHIRVHTGLKPYACTQCRKSFHTRGNCILHGKIHTGETPYLCGQCGKSFTQRGSLAVHQRSCSQRLTL
RUO
ZNF32
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7580
ZNF32 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KOX30
ZNF32
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers