missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF26 Partial ORF (NP_062537, 445 a.a. - 533 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007574-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107725

  • 349.00€ / 10µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

Sequence: QRTHSTEKPYECNECEKAYPRKASLQIHQKTHSGEKPFKCSECGKAFTQKSSLSEHQRVHTGEKPWKCSECGKSFCWNSGLRIHRKTHK
Specifications

Specifications

NP_062537
Liquid
7574
ZNF26 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20755/KOX20
ZNF26
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QRTHSTEKPYECNECEKAYPRKASLQIHQKTHSGEKPFKCSECGKAFTQKSSLSEHQRVHTGEKPWKCSECGKSFCWNSGLRIHRKTHK
RUO
ZNF26
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers