Learn More
Abnova™ Human ZNF16 Partial ORF (NP_008889, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007564-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a C2H2 type of zinc finger, and thus may function as a transcription factor. This gene is located in a region close to ZNF7/KOX4, a gene also encoding a zinc finger protein, on chromosome 8. Two alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: PSLRTRREEAEMELSVPGPSPWTPAAQARVRDAPAVTHPGSAACGTPCCSDTELEAICPHYQQPDCDTRTEDKEFLHKEDIHEDLESQAEISENYAGDVSpecifications
NP_008889 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PSLRTRREEAEMELSVPGPSPWTPAAQARVRDAPAVTHPGSAACGTPCCSDTELEAICPHYQQPDCDTRTEDKEFLHKEDIHEDLESQAEISENYAGDV | |
RUO | |
ZNF16 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7564 | |
ZNF16 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KOX9 | |
ZNF16 | |
Recombinant | |
wheat germ expression system |