missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF16 Partial ORF (NP_008889, 2 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007564-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157705

  • 335.00€ / 10µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

The protein encoded by this gene contains a C2H2 type of zinc finger, and thus may function as a transcription factor. This gene is located in a region close to ZNF7/KOX4, a gene also encoding a zinc finger protein, on chromosome 8. Two alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]

Sequence: PSLRTRREEAEMELSVPGPSPWTPAAQARVRDAPAVTHPGSAACGTPCCSDTELEAICPHYQQPDCDTRTEDKEFLHKEDIHEDLESQAEISENYAGDV
Specifications

Specifications

NP_008889
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PSLRTRREEAEMELSVPGPSPWTPAAQARVRDAPAVTHPGSAACGTPCCSDTELEAICPHYQQPDCDTRTEDKEFLHKEDIHEDLESQAEISENYAGDV
RUO
ZNF16
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7564
ZNF16 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KOX9
ZNF16
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers