missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF134 Partial ORF (NP_003426.2, 105 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007693-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107825

  • 349.00€ / 10µg

Estimated Shipment: 11-07-2023
to see stock.
Add to basket

Description

Description

Sequence: PLRRDKSEASIVRNCTVSKEPHPSEKPFTCKEEQKNFQATLGGCQQKAIHSKRKTHRSTESGDAFHGEQMHYKCSECGKAFSRKDTLVQ
Specifications

Specifications

NP_003426.2
Liquid
7693
ZNF134 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC138499/MGC141970/pHZ-15
ZNF134
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PLRRDKSEASIVRNCTVSKEPHPSEKPFTCKEEQKNFQATLGGCQQKAIHSKRKTHRSTESGDAFHGEQMHYKCSECGKAFSRKDTLVQ
RUO
ZNF134
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers