Learn More
Abnova™ Human ZNF12 Partial ORF (NP_057349, 69 a.a. - 175 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007559-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the krueppel C2H2-type zinc-finger protein family and encodes a protein with eight C2H2-type zinc fingers and a KRAB domain. This nuclear protein is involved in developmental control of gene expression. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: VEGEFLLQSYPDEVWQTDDLIERIQEEENKPSRQTVFIETLIEERGNVPGKTFDVETNPVPSRKIAYKNSLCDSCEKCLTSVSEYISSDGSYARMKADECSGCGKSLSpecifications
NP_057349 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VEGEFLLQSYPDEVWQTDDLIERIQEEENKPSRQTVFIETLIEERGNVPGKTFDVETNPVPSRKIAYKNSLCDSCEKCLTSVSEYISSDGSYARMKADECSGCGKSL | |
RUO | |
ZNF12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7559 | |
ZNF12 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GIOT-3/HZF11/KOX3/ZNF325 | |
ZNF12 | |
Recombinant | |
wheat germ expression system |