missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF12 Partial ORF (NP_057349, 69 a.a. - 175 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007559-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107705

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene is a member of the krueppel C2H2-type zinc-finger protein family and encodes a protein with eight C2H2-type zinc fingers and a KRAB domain. This nuclear protein is involved in developmental control of gene expression. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: VEGEFLLQSYPDEVWQTDDLIERIQEEENKPSRQTVFIETLIEERGNVPGKTFDVETNPVPSRKIAYKNSLCDSCEKCLTSVSEYISSDGSYARMKADECSGCGKSL
Specifications

Specifications

NP_057349
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.51kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VEGEFLLQSYPDEVWQTDDLIERIQEEENKPSRQTVFIETLIEERGNVPGKTFDVETNPVPSRKIAYKNSLCDSCEKCLTSVSEYISSDGSYARMKADECSGCGKSL
RUO
ZNF12
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7559
ZNF12 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GIOT-3/HZF11/KOX3/ZNF325
ZNF12
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers