missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZMYM3 Full-length ORF (AAH13009.1, 1 a.a. - 495 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

349.00€ - 528.00€

Specifications

Accession Number AAH13009.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9203
Molecular Weight (g/mol) 78.9kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16103392
View Documents Promotion Details
Abnova™
H00009203-P01.10UG
10 ug
349.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
16113392
View Documents Promotion Details
Abnova™
H00009203-P01.25UG
25 ug
528.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
Description

Description

Sequence: MDPSDFPSPFDPLTLPEKPLAGDLPVDMEFGEDLLESQTAPTRGWAPPGPSPSSGALDLLDTPAGLEKDPGVLDGATELLGLGGLLYKAPSPPEVDHGPEGTLAWDAGDQTLEPGPGGQTPEVVPPDPGAGANSCSPEGLLEPLAPDSPITLQSPHIEEEETTSIATARRGSPGQEEELPQGQPQSPNAPPSPSVGETLGDGINSSQTKPGGSSPPAHPSLPGDGLTAKASEKPPERKRSERVRRAEPPKPEVVDSTESIPVSDEDSDAMVDDPNDEDFVPFRPRRSPRMSLRSSVSQRAGRSAVGTKMTCAHCRTPLQKGQTAYQRKGLPQLFCSSSCLTTFSKKPSGKKTCTFCKKEIWNTKDSVVAQTGSGGSFHEFCTSVCLSLYEAQQQRPIPQSGDPADATRCSICQKTGEVLHEVSNGSVVHRLCSDSCFSKFRANKGLKTNCCDQCGAYIYTKTGSPGPELLFHEGQQKRFCNTTCLGAYKKVGPRE
Specifications

Specifications

AAH13009.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
78.9kDa
Glutathione Sepharose 4 Fast Flow
MDPSDFPSPFDPLTLPEKPLAGDLPVDMEFGEDLLESQTAPTRGWAPPGPSPSSGALDLLDTPAGLEKDPGVLDGATELLGLGGLLYKAPSPPEVDHGPEGTLAWDAGDQTLEPGPGGQTPEVVPPDPGAGANSCSPEGLLEPLAPDSPITLQSPHIEEEETTSIATARRGSPGQEEELPQGQPQSPNAPPSPSVGETLGDGINSSQTKPGGSSPPAHPSLPGDGLTAKASEKPPERKRSERVRRAEPPKPEVVDSTESIPVSDEDSDAMVDDPNDEDFVPFRPRRSPRMSLRSSVSQRAGRSAVGTKMTCAHCRTPLQKGQTAYQRKGLPQLFCSSSCLTTFSKKPSGKKTCTFCKKEIWNTKDSVVAQTGSGGSFHEFCTSVCLSLYEAQQQRPIPQSGDPADATRCSICQKTGEVLHEVSNGSVVHRLCSDSCFSKFRANKGLKTNCCDQCGAYIYTKTGSPGPELLFHEGQQKRFCNTTCLGAYKKVGPRE
RUO
ZMYM3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
9203
ZMYM3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DXS6673E/KIAA0385/MYM/XFIM/ZNF198L2/ZNF261
ZMYM3
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications