missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZMAT5 Full-length ORF (NP_001003692.1, 1 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055954-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16101993

  • 349.00€ / 10µg

Estimated Shipment: 18-07-2023
to see stock.
Add to basket

Description

Description

Sequence: MGKRYFCDYCDRSFQDNLHNRKKHLNGLQHLKAKKVWYDMFRDAAAILLDEQNKRPCRKFLLTGQCDFGSNCRFSHMSERDLQELSIQVEEERRAREWLLDAPELPEGHLEDWLEKRAKRLSSAPSSRAEPIRTTVFQYPVGWPPVQELPPSLRAPPPGGWPLQPRVQWG
Specifications

Specifications

NP_001003692.1
Liquid
55954
ZMAT5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGKRYFCDYCDRSFQDNLHNRKKHLNGLQHLKAKKVWYDMFRDAAAILLDEQNKRPCRKFLLTGQCDFGSNCRFSHMSERDLQELSIQVEEERRAREWLLDAPELPEGHLEDWLEKRAKRLSSAPSSRAEPIRTTVFQYPVGWPPVQELPPSLRAPPPGGWPLQPRVQWG
RUO
ZMAT5
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ZMAT5
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers