missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZFPM1 Partial ORF (NP_722520, 86 a.a. - 195 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00161882-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192247

  • 349.00€ / 10µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

Sequence: WSGPDELEPVVQDGQRRIRARLSLATGLSWGPFHGSVQTRASSPRQAEPSPALTLLLVDEACWLRTLPQALTEAEANTEIHRKDDALWCRVTKPVPAGGLLSVLLTAEPH
Specifications

Specifications

NP_722520
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
WSGPDELEPVVQDGQRRIRARLSLATGLSWGPFHGSVQTRASSPRQAEPSPALTLLLVDEACWLRTLPQALTEAEANTEIHRKDDALWCRVTKPVPAGGLLSVLLTAEPH
RUO
ZFPM1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
161882
ZFPM1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FOG/FOG1/ZNF408/ZNF89A
ZFPM1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers