missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZFP37 Partial ORF (NP_003399.1, 193 a.a. - 288 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007539-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157655

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Sequence: HSRNCVKRKSDAAKEHKKSFNHSLSDTRKGKKQTGKKHEKLSSHSSSDKCNKTGKKHDKLCCHSSSHIKQDKIQTGEKHEKSPSLSSSTKHEKPQA
Specifications

Specifications

NP_003399.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HSRNCVKRKSDAAKEHKKSFNHSLSDTRKGKKQTGKKHEKLSSHSSSDKCNKTGKKHDKLCCHSSSHIKQDKIQTGEKHEKSPSLSSSTKHEKPQA
RUO
ZFP37
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7539
ZFP37 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ38524
ZFP37
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers