missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZFP161 Partial ORF (NP_003400.2, 311 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007541-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107665

  • 349.00€ / 10µg

Estimated Shipment: 17-07-2023
to see stock.
Add to basket

Description

Description

Sequence: TKGFTTQAHLKEHLKIHTGYKPYSCEVCGKSFIRAPDLKKHERVHSNERPFACHMCDKAFKHKSHLKDHERRHRGEKPFVCGSCTKAFAKASDLKRHENNMHSERKQVTP
Specifications

Specifications

NP_003400.2
Liquid
7541
ZFP161 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126126/ZBTB14/ZF5/ZNF478
ZFP161
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TKGFTTQAHLKEHLKIHTGYKPYSCEVCGKSFIRAPDLKKHERVHSNERPFACHMCDKAFKHKSHLKDHERRHRGEKPFVCGSCTKAFAKASDLKRHENNMHSERKQVTP
RUO
ZFP161
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers