missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZC3HAV1L Full-length ORF (NP_542391.1, 1 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00092092-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MAEPTVCSFLTKVLCAHGGRMFLKDLRGHVELSEARLRDVLQRAGPERFLLQEVETQEGLGDAEAEAAAGAVGGGGTSAWRVVAVSSVRLCARYQRGECQACDQLHFCRRHMLGKCPNRDCWSTCTLSHDIHTPVNMQVLKSHGLFGLNENQLRILLLQNDPCLLPEVCLLYNKGEALYGYCNLKDKCNKFHVCKSFVKGECKLQTCKRSHQLIHAASLKLLQDQGLNIPSVVNFQIISTYKHIKLHKMLENTDNSSPSTEHSQGLEKQGVHAAGAAEAGPLASVPAQSAKKPCPVSCEKSpecifications
NP_542391.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C7orf39/MGC14289 | |
ZC3HAV1L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
92092 | |
ZC3HAV1L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEPTVCSFLTKVLCAHGGRMFLKDLRGHVELSEARLRDVLQRAGPERFLLQEVETQEGLGDAEAEAAAGAVGGGGTSAWRVVAVSSVRLCARYQRGECQACDQLHFCRRHMLGKCPNRDCWSTCTLSHDIHTPVNMQVLKSHGLFGLNENQLRILLLQNDPCLLPEVCLLYNKGEALYGYCNLKDKCNKFHVCKSFVKGECKLQTCKRSHQLIHAASLKLLQDQGLNIPSVVNFQIISTYKHIKLHKMLENTDNSSPSTEHSQGLEKQGVHAAGAAEAGPLASVPAQSAKKPCPVSCEK | |
RUO | |
ZC3HAV1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |