missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZC3H12A Full-length ORF (BAB15581.1, 1 a.a. - 599 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080149-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16164783

  • 359.00€ / 10µg

Estimated Shipment: 22-08-2022
to see stock.
Add to basket

Description

Description

ZC3H12A is an MCP1 (CCL2; MIM 158105)-induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis.[supplied by OMIM]

Sequence: MSGPCGEKPVLEASPTMSLWEFEDSHSRQGTPRPGQELAAEEASALELQMKVDFFRKLGYSSTEIHSVLQKLGVQADTNTVLGELVKHGTATERERQTSPDPCPQLPLVPRGGGTPKAPNLEPPLPEEEKEGSDLRPVVIDGSNVAMSHGNKEVFSCRGILLAVNWFLERGHTDITVFVPSWRKEQPRPDVPITDQHILRELEKKKILVFTPSRRVGGKRVVCYDDRFIVKLAYESDGIVVSNDTYRDLQGERQEWKRFIEERLLMYSFVNDKFMPPDDPLGRHGPSLDNFLRKKPLTLEHRKQPCPYGRKCTYGIKCRFFHPERPSCPQRSVADELRANALLSPPRAPSKDKNGRRPSPSSQSSSLLTESEQCSLDGKKLGAQASPGSRQEGLTQTYAPSGRSLAPSGGSGSSFGPTDWLPQTLDSLPYVSQDCLDSGIGSLESQMSELWGVRGGGPGEPGPPRAPYTGYSPYGSELPATAAFSAFGRAMGAGHFSVPADYPPAPPAFPPREYWSEPYPLPPPTSVLQEPPVQSPGAGRSPWGRADSLAKEQASVYTKLCGVFPPHLVEAVMGRFPQLLDPQQLAAEILSYKSQHPSE
Specifications

Specifications

BAB15581.1
Liquid
80149
ZC3H12A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MSGPCGEKPVLEASPTMSLWEFEDSHSRQGTPRPGQELAAEEASALELQMKVDFFRKLGYSSTEIHSVLQKLGVQADTNTVLGELVKHGTATERERQTSPDPCPQLPLVPRGGGTPKAPNLEPPLPEEEKEGSDLRPVVIDGSNVAMSHGNKEVFSCRGILLAVNWFLERGHTDITVFVPSWRKEQPRPDVPITDQHILRELEKKKILVFTPSRRVGGKRVVCYDDRFIVKLAYESDGIVVSNDTYRDLQGERQEWKRFIEERLLMYSFVNDKFMPPDDPLGRHGPSLDNFLRKKPLTLEHRKQPCPYGRKCTYGIKCRFFHPERPSCPQRSVADELRANALLSPPRAPSKDKNGRRPSPSSQSSSLLTESEQCSLDGKKLGAQASPGSRQEGLTQTYAPSGRSLAPSGGSGSSFGPTDWLPQTLDSLPYVSQDCLDSGIGSLESQMSELWGVRGGGPGEPGPPRAPYTGYSPYGSELPATAAFSAFGRAMGAGHFSVPADYPPAPPAFPPREYWSEPYPLPPPTSVLQEPPVQSPGAGRSPWGRADSLAKEQASVYTKLCGVFPPHLVEAVMGRFPQLLDPQQLAAEILSYKSQHPSE
RUO
ZC3H12A
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ23231/MCPIP/MCPIP1/RP3-423B22.1/dJ423B22.1
ZC3H12A
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers