Learn More
Abnova™ Human XG Full-length ORF (AAI00768.1, 1 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAI00768.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7499 |
Molecular Weight (g/mol) | 46.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16171892
|
Abnova™
H00007499-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16161892
|
Abnova™
H00007499-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
This gene encodes the XG blood group antigen, and is located at the pseudoautosomal boundary on the short (p) arm of chromosome X. The three 5' exons reside in the pseudoautosomal region and the remaining exons within the X-specific end. A truncated copy of this gene is found on the Y chromosome at the pseudoautosomal boundary. It is transcribed, but not expected to make a Y-chromosome specific gene product. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MESWWGLPCLAFLCFLMHARGQRDFDLADALDDPEPTKKPNSDIYPKPKPPYYPQPENPDSGGNIYPRPKPRPQPQPGNSGNSGGSYFNDVDRDDGRYPPRPRPRPPAGGGGGGYSSYGNSDNTHGGDHHSTYGNPEGNMVAKIVSPIVSVVVVTLLGAAASYFKLNNRRNCFRTHEPENVSpecifications
AAI00768.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MESWWGLPCLAFLCFLMHARGQRDFDLADALDDPEPTKKPNSDIYPKPKPPYYPQPENPDSGGNIYPRPKPRPQPQPGNSGNSGGSYFNDVDRDDGRYPPRPRPRPPAGGGGGGYSSYGNSDNTHGGDHHSTYGNPEGNMVAKIVSPIVSVVVVTLLGAAASYFKLNNRRNCFRTHEPENV | |
RUO | |
XG | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7499 | |
XG (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC118758/MGC118759/MGC118760/MGC118761/PBDX | |
XG | |
Recombinant | |
wheat germ expression system |