missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human XAGE2 Full-length ORF (NP_570133.1, 1 a.a. - 111 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009502-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193742

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in normal testis, and in Ewing's sarcoma, rhabdomyosarcoma, a breast cancer and a germ cell tumor. The protein encoded by this gene shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens. [provided by RefSeq]

Sequence: MSWRGRSTYRPRPRRSLQPPELIGAMLEPTDEEPKEEKPPTKSRNPTPDQKREDDQGAAEIQVPDLEADLQELCQTKTGDGCEGGTDVKGKILPKAEHFKMPEAGEGKSQV
Specifications

Specifications

NP_570133.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GAGED3/XAGE-2
XAGE2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9502
XAGE2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSWRGRSTYRPRPRRSLQPPELIGAMLEPTDEEPKEEKPPTKSRNPTPDQKREDDQGAAEIQVPDLEADLQELCQTKTGDGCEGGTDVKGKILPKAEHFKMPEAGEGKSQV
RUO
XAGE2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers