missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human XAGE1 Full-length ORF (NP_597673.1, 1 a.a. - 69 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009503-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123752

  • 359.00€ / 10µg

Estimated Shipment: 07-10-2022
to see stock.
Add to basket

Description

Description

This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in Ewing's sarcoma, alveolar rhabdomyosarcoma and normal testis. The protein encoded by this gene contains a nuclear localization signal and shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens. Alternative splicing of this gene, in addition to the use of an alternative transcription start site and an alternative translation initiation codon, results in multiple variants that encode different isoforms. [provided by RefSeq]

Sequence: MESPKKKNQQLKVGILHLGSRQKKIRIQLRSQVLGREMRDMEGDLQELHQSNTGDKSGFGFRRQGEDNT
Specifications

Specifications

NP_597673.1
Liquid
9503
XAGE1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MESPKKKNQQLKVGILHLGSRQKKIRIQLRSQVLGREMRDMEGDLQELHQSNTGDKSGFGFRRQGEDNT
RUO
XAGE1D
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
XAGE1D
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers