Learn More
Abnova™ Human XAGE1 Full-length ORF (NP_597673.1, 1 a.a. - 69 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009503-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in Ewing's sarcoma, alveolar rhabdomyosarcoma and normal testis. The protein encoded by this gene contains a nuclear localization signal and shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens. Alternative splicing of this gene, in addition to the use of an alternative transcription start site and an alternative translation initiation codon, results in multiple variants that encode different isoforms. [provided by RefSeq]
Sequence: MESPKKKNQQLKVGILHLGSRQKKIRIQLRSQVLGREMRDMEGDLQELHQSNTGDKSGFGFRRQGEDNTSpecifications
NP_597673.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
XAGE1D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9503 | |
XAGE1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MESPKKKNQQLKVGILHLGSRQKKIRIQLRSQVLGREMRDMEGDLQELHQSNTGDKSGFGFRRQGEDNT | |
RUO | |
XAGE1D | |
Recombinant | |
wheat germ expression system |