Learn More
Abnova™ Human WIPI1 Full-length ORF (NP_060453.2, 1 a.a. - 446 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055062-P01.25ug
Additional Details : Weight : 0.00010kg
Description
WD40 repeat proteins are key components of many essential biologic functions. They regulate the assembly of multiprotein complexes by presenting a beta-propeller platform for simultaneous and reversible protein-protein interactions. Members of the WIPI subfamily of WD40 repeat proteins, such as WIPI1, have a 7-bladed propeller structure and contain a conserved motif for interaction with phospholipids (Proikas-Cezanne et al., 2004 [PubMed 15602573]).[supplied by OMIM]
Sequence: MEAEAADAPPGGVESALSCFSFNQDCTSLAIGTKAGYKLFSLSSVEQLDQVHGSNEIPDVYIVERLFSSSLVVVVSHTKPRQMNVYHFKKGTEICNYSYSSNILSIRLNRQRLLVCLEESIYIHNIKDMKLLKTLLDIPANPTGLCALSINHSNSYLAYPGSLTSGEIVLYDGNSLKTVCTIAAHEGTLAAITFNASGSKLASASEKGTVIRVFSVPDGQKLYEFRRGMKRYVTISSLVFSMDSQFLCASSNTETVHIFKLEQVTNSRPEEPSTWSGYMGKMFMAATNYLPTQVSDMMHQDRAFATARLNFSGQRNICTLSTIQKLPRLLVASSSGHLYMYNLDPQDGGECVLIKTHSLLGSGTTEENKENDLRPSLPQSYAATVARPSASSASTVPGYSEDGGALRGEVIPEHEFATGPVCLDDENEFPPIILCRGNQKGKTKQSSpecifications
NP_060453.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATG18/FLJ10055/WIPI49 | |
WIPI1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55062 | |
WIPI1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEAEAADAPPGGVESALSCFSFNQDCTSLAIGTKAGYKLFSLSSVEQLDQVHGSNEIPDVYIVERLFSSSLVVVVSHTKPRQMNVYHFKKGTEICNYSYSSNILSIRLNRQRLLVCLEESIYIHNIKDMKLLKTLLDIPANPTGLCALSINHSNSYLAYPGSLTSGEIVLYDGNSLKTVCTIAAHEGTLAAITFNASGSKLASASEKGTVIRVFSVPDGQKLYEFRRGMKRYVTISSLVFSMDSQFLCASSNTETVHIFKLEQVTNSRPEEPSTWSGYMGKMFMAATNYLPTQVSDMMHQDRAFATARLNFSGQRNICTLSTIQKLPRLLVASSSGHLYMYNLDPQDGGECVLIKTHSLLGSGTTEENKENDLRPSLPQSYAATVARPSASSASTVPGYSEDGGALRGEVIPEHEFATGPVCLDDENEFPPIILCRGNQKGKTKQS | |
RUO | |
WIPI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |