Learn More
Abnova™ Human WFIKKN2 Full-length ORF (NP_783165.1, 1 a.a. - 576 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00124857-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The WFIKKN1 protein contains a WAP domain, follistatin domain, immunoglobulin domain, two tandem Kunitz domains, and an NTR domain. This gene encodes a WFIKKN1-related protein which has the same domain organization as the WFIKKN1 protein. The WAP-type, follistatin type, Kunitz-type, and NTR-type protease inhibitory domains may control the action of multiple types of proteases. [provided by RefSeq]
Sequence: MWAPRCRRFWSRWEQVAALLLLLLLLGVPPRSLALPPIRYSHAGICPNDMNPNLWVDAQSTCRRECETDQECETYEKCCPNVCGTKSCVAARYMDVKGKKGPVGMPKEATCDHFMCLQQGSECDIWDGQPVCKCKDRCEKEPSFTCASDGLTYYNRCYMDAEACSKGITLAVVTCRYHFTWPNTSPPPPETTMHPTTASPETPELDMAAPALLNNPVHQSVTMGETVSFLCDVVGRPRPEITWEKQLEDRENVVMRPNHVRGNVVVTNIAQLVIYNAQLQDAGIYTCTARNVAGVLRADFPLSVVRGHQAAATSESSPNGTAFPAAECLKPPDSEDCGEEQTRWHFDAQANNCLTFTFGHCHRNLNHFETYEACMLACMSGPLAACSLPALQGPCKAYAPRWAYNSQTGQCQSFVYGGCEGNGNNFESREACEESCPFPRGNQRCRACKPRQKLVTSFCRSDFVILGRVSELTEEPDSGRALVTVDEVLKDEKMGLKFLGQEPLEVTLLHVDWACPCPNVTVSEMPLIIMGEVDGGMAMLRPDSFVGASSARRVRKLREVMHKKTCDVLKEFLGLHSpecifications
NP_783165.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
90.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WFIKKNRP | |
WFIKKN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
124857 | |
WFIKKN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWAPRCRRFWSRWEQVAALLLLLLLLGVPPRSLALPPIRYSHAGICPNDMNPNLWVDAQSTCRRECETDQECETYEKCCPNVCGTKSCVAARYMDVKGKKGPVGMPKEATCDHFMCLQQGSECDIWDGQPVCKCKDRCEKEPSFTCASDGLTYYNRCYMDAEACSKGITLAVVTCRYHFTWPNTSPPPPETTMHPTTASPETPELDMAAPALLNNPVHQSVTMGETVSFLCDVVGRPRPEITWEKQLEDRENVVMRPNHVRGNVVVTNIAQLVIYNAQLQDAGIYTCTARNVAGVLRADFPLSVVRGHQAAATSESSPNGTAFPAAECLKPPDSEDCGEEQTRWHFDAQANNCLTFTFGHCHRNLNHFETYEACMLACMSGPLAACSLPALQGPCKAYAPRWAYNSQTGQCQSFVYGGCEGNGNNFESREACEESCPFPRGNQRCRACKPRQKLVTSFCRSDFVILGRVSELTEEPDSGRALVTVDEVLKDEKMGLKFLGQEPLEVTLLHVDWACPCPNVTVSEMPLIIMGEVDGGMAMLRPDSFVGASSARRVRKLREVMHKKTCDVLKEFLGLH | |
RUO | |
WFIKKN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |