missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WFIKKN2 Full-length ORF (NP_783165.1, 1 a.a. - 576 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00124857-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107643

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

The WFIKKN1 protein contains a WAP domain, follistatin domain, immunoglobulin domain, two tandem Kunitz domains, and an NTR domain. This gene encodes a WFIKKN1-related protein which has the same domain organization as the WFIKKN1 protein. The WAP-type, follistatin type, Kunitz-type, and NTR-type protease inhibitory domains may control the action of multiple types of proteases. [provided by RefSeq]

Sequence: MWAPRCRRFWSRWEQVAALLLLLLLLGVPPRSLALPPIRYSHAGICPNDMNPNLWVDAQSTCRRECETDQECETYEKCCPNVCGTKSCVAARYMDVKGKKGPVGMPKEATCDHFMCLQQGSECDIWDGQPVCKCKDRCEKEPSFTCASDGLTYYNRCYMDAEACSKGITLAVVTCRYHFTWPNTSPPPPETTMHPTTASPETPELDMAAPALLNNPVHQSVTMGETVSFLCDVVGRPRPEITWEKQLEDRENVVMRPNHVRGNVVVTNIAQLVIYNAQLQDAGIYTCTARNVAGVLRADFPLSVVRGHQAAATSESSPNGTAFPAAECLKPPDSEDCGEEQTRWHFDAQANNCLTFTFGHCHRNLNHFETYEACMLACMSGPLAACSLPALQGPCKAYAPRWAYNSQTGQCQSFVYGGCEGNGNNFESREACEESCPFPRGNQRCRACKPRQKLVTSFCRSDFVILGRVSELTEEPDSGRALVTVDEVLKDEKMGLKFLGQEPLEVTLLHVDWACPCPNVTVSEMPLIIMGEVDGGMAMLRPDSFVGASSARRVRKLREVMHKKTCDVLKEFLGLH
Specifications

Specifications

NP_783165.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
90.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
WFIKKNRP
WFIKKN2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
124857
WFIKKN2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWAPRCRRFWSRWEQVAALLLLLLLLGVPPRSLALPPIRYSHAGICPNDMNPNLWVDAQSTCRRECETDQECETYEKCCPNVCGTKSCVAARYMDVKGKKGPVGMPKEATCDHFMCLQQGSECDIWDGQPVCKCKDRCEKEPSFTCASDGLTYYNRCYMDAEACSKGITLAVVTCRYHFTWPNTSPPPPETTMHPTTASPETPELDMAAPALLNNPVHQSVTMGETVSFLCDVVGRPRPEITWEKQLEDRENVVMRPNHVRGNVVVTNIAQLVIYNAQLQDAGIYTCTARNVAGVLRADFPLSVVRGHQAAATSESSPNGTAFPAAECLKPPDSEDCGEEQTRWHFDAQANNCLTFTFGHCHRNLNHFETYEACMLACMSGPLAACSLPALQGPCKAYAPRWAYNSQTGQCQSFVYGGCEGNGNNFESREACEESCPFPRGNQRCRACKPRQKLVTSFCRSDFVILGRVSELTEEPDSGRALVTVDEVLKDEKMGLKFLGQEPLEVTLLHVDWACPCPNVTVSEMPLIIMGEVDGGMAMLRPDSFVGASSARRVRKLREVMHKKTCDVLKEFLGLH
RUO
WFIKKN2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers