Learn More
Abnova™ Human WDR19 Full-length ORF (AAH32578.1, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057728-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains six WD repeats, a clathrin heavy-chain repeat, and three transmembrane domains. This gene is conserved from C. elegans to human. It may participate in androgen-regulated signaling mechanisms or in the vesicular trafficking of androgen-regulated secretory processes. Alternatively spliced transcript variants encoding distinct isoforms have been reported but the full-length nature of one of these variants has not been defined. [provided by RefSeq]
Sequence: MKRIFSLLEKTWLGAPIQFAWQKTSGNYLAVTGADYIVKIFDRHGQKRSEINLPGNCVAMDWDKDGDVLAVIAEKSSCIYLWDANTNKTSQLDNGMRDQMSFLLWSKVGSFLAVGTVKGNLLIYNHQTSRKIPVLGKHTKRITCGCWNAENLLALGGEDKMITVSNQEGDTIRQTQVRSEPSNMQFFLMKMDDRTSAAESMISVVLGKKTLFFLNLNEPDNPADLEFQQDFGNIVCYNWYGDGRIMIGFSCGHFVVISTHTGELGQEIFQARNHKDNLTSIAVSQTLNKVATCGDNCIKIQDLVDLKDMYVILNLDEENKGLGTLSWTDDGQLLALSTQRGSLHVFLTKLPILGDACSTRIAYLTSLLEVTVANPVEGELPITVSVDVEPNFVAVGLYHLAVGMNNRAWFYVLGENAVKKIERYGVSGNSSQYLPSFSpecifications
AAH32578.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23127/KIAA1638/ORF26/PWDMP | |
WDR19 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
57728 | |
WDR19 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKRIFSLLEKTWLGAPIQFAWQKTSGNYLAVTGADYIVKIFDRHGQKRSEINLPGNCVAMDWDKDGDVLAVIAEKSSCIYLWDANTNKTSQLDNGMRDQMSFLLWSKVGSFLAVGTVKGNLLIYNHQTSRKIPVLGKHTKRITCGCWNAENLLALGGEDKMITVSNQEGDTIRQTQVRSEPSNMQFFLMKMDDRTSAAESMISVVLGKKTLFFLNLNEPDNPADLEFQQDFGNIVCYNWYGDGRIMIGFSCGHFVVISTHTGELGQEIFQARNHKDNLTSIAVSQTLNKVATCGDNCIKIQDLVDLKDMYVILNLDEENKGLGTLSWTDDGQLLALSTQRGSLHVFLTKLPILGDACSTRIAYLTSLLEVTVANPVEGELPITVSVDVEPNFVAVGLYHLAVGMNNRAWFYVLGENAVKKIERYGVSGNSSQYLPSF | |
RUO | |
WDR19 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |