Learn More
ACROBiosystems Human VSIG4 Protein, His Tag
Brand: ACROBiosystems VS4-H5226-100UG
Additional Details : Weight : 0.00001kg
Description
V-set and immunoglobulin domain-containing protein 4 (VSIG4) is also known as Protein Z39Ig, is a type I transmembrane glycoprotein. VSIG4 is a B7 family-related protein and an Ig superfamily member. VSIG4 contains two Ig-like (immunoglobulin-like) domains. VSIG4 is abundantly expressed in several fetal tissues. In adult tissues, the highest expression of VSIG4 is in lung and placenta. VSIG4 functions as a negative regulator of T cell activation, and may be involved in the maintenance of peripheral T cell tolerance, and is also identified as a potent suppressor of established inflammation. VSIG4 is a phagocytic receptor, strong negative regulator of T-cell proliferation and IL2 production.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.1 kDa. The protein migrates as 35-47 kDa under reducing (R) condition (SDS-PAGE).Specifications
AAH10525 | |
100 μg | |
Arg 20 - Pro 283 | |
VSIG4,CRIg,Z39IG | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
30.1 kDa | |
Human VSIG4, His Tag (VS4-H5226) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Pro 283 (Accession # AAH10525). | |
-20°C | |
VSIG4 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |