Learn More
ACROBiosystems Human VSIG2 Protein, His Tag
Brand: ACROBiosystems VS2-H5225-1MG
Additional Details : Weight : 0.00001kg
Description
V-set and immunoglobulin domain-containing protein 2 (VSIG2) is also known as Cortical thymocyte-like protein, CTH, CTXL, is a single-pass type I membrane protein which contains Ig-like C2-type (immunoglobulin-like) domain and one Ig-like V-type (immunoglobulin-like) domain. V-set domains are Ig-like domains resembling the antibody variable domain. VSIG2 is highly expressed in stomach, colon, prostate, trachea and thyroid glands and weakly in bladder and lung. V-set domains are found in diverse protein families, including immunoglobulin light and heavy chains.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.4 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH07313 | |
1 mg | |
Val 24 - Ala 243 | |
VSIG2,CT-like protein,CTH,CTXL | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
24.4 kDa | |
Human VSIG2, His Tag (VS2-H5225) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Ala 243 (Accession # AAH07313). | |
-20°C | |
VSIG2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |