Learn More
Abnova™ Human VPS33B Partial ORF (NP_061138.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026276-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and encodes the human ortholog of rat Vps33b which is homologous to the yeast class C Vps33 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. [provided by RefSeq]
Sequence: MAFPHRPDAPELPDFSMLKRLARDQLIYLLEQLPGKKDLFIEADLMSPLDRIANVSILKQHEVDKLYKVENKPALSSNEQLCFLVRPRIKNMRYIASLVNSpecifications
NP_061138.2 | |
Liquid | |
26276 | |
VPS33B (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14848 | |
VPS33B | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAFPHRPDAPELPDFSMLKRLARDQLIYLLEQLPGKKDLFIEADLMSPLDRIANVSILKQHEVDKLYKVENKPALSSNEQLCFLVRPRIKNMRYIASLVN | |
RUO | |
VPS33B | |
Wheat Germ (in vitro) | |
GST |