missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human VPS33A Full-length ORF (NP_075067.2, 1 a.a. - 596 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00065082-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16103883

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and it encodes a protein similar to the yeast class C Vps33 protein. The mammalian class C VPS proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. [provided by RefSeq]

Sequence: MAAHLSYGRVNLNVLREAVRRELREFLDKCAGSKAIVWDEYLTGPFGLIAQYSLLKEHEVEKMFTLKGNRLPAADVKNIIFFVRPRLELMDIIAENVLSEDRRGPTRDFHILFVPRRSLLCEQRLKDLGVLGSFIHREEYSLDLIPFDGDLLSMESEGAFKECYLEGDQTSLYHAAKGLMTLQALYGTIPQIFGKGECARQVANMMIRMKREFTGSQNSIFPVFDNLLLLDRNVDLLTPLATQLTYEGLIDEIYGIQNSYVKLPPEKFAPKKQGDGGKDLPTEAKKLQLNSAEELYAEIRDKNFNAVGSVLSKKAKIISAAFEERHNAKTVGEIKQFVSQLPHMQAARGSLANHTSIAELIKDVTTSEDFFDKLTVEQEFMSGIDTDKVNNYIEDCIAQKHSLIKVLRLVCLQSVCNSGLKQKVLDYYKREILQTYGYEHILTLHNLEKAGLLKPQTGGRNNYPTIRKTLRLWMDDVNEQNPTDISYVYSGYAPLSVRLAQLLSRPGWRSIEEVLRILPGPHFEERQPLPTGLQKKRQPGENRVTLIFFLGGVTFAEIAALRFLSQLEDGGTEYVIATTKLMNGTSWIEALMEKPF
Specifications

Specifications

NP_075067.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
94kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ22395/FLJ23187
VPS33A
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
65082
VPS33A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAHLSYGRVNLNVLREAVRRELREFLDKCAGSKAIVWDEYLTGPFGLIAQYSLLKEHEVEKMFTLKGNRLPAADVKNIIFFVRPRLELMDIIAENVLSEDRRGPTRDFHILFVPRRSLLCEQRLKDLGVLGSFIHREEYSLDLIPFDGDLLSMESEGAFKECYLEGDQTSLYHAAKGLMTLQALYGTIPQIFGKGECARQVANMMIRMKREFTGSQNSIFPVFDNLLLLDRNVDLLTPLATQLTYEGLIDEIYGIQNSYVKLPPEKFAPKKQGDGGKDLPTEAKKLQLNSAEELYAEIRDKNFNAVGSVLSKKAKIISAAFEERHNAKTVGEIKQFVSQLPHMQAARGSLANHTSIAELIKDVTTSEDFFDKLTVEQEFMSGIDTDKVNNYIEDCIAQKHSLIKVLRLVCLQSVCNSGLKQKVLDYYKREILQTYGYEHILTLHNLEKAGLLKPQTGGRNNYPTIRKTLRLWMDDVNEQNPTDISYVYSGYAPLSVRLAQLLSRPGWRSIEEVLRILPGPHFEERQPLPTGLQKKRQPGENRVTLIFFLGGVTFAEIAALRFLSQLEDGGTEYVIATTKLMNGTSWIEALMEKPF
RUO
VPS33A
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers