Learn More
Abnova™ Human VMD2L3 Partial ORF (NP_689652, 169 a.a. - 267 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00144453-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
BEST3 belongs to the bestrophin family of anion channels, which includes BEST1 (MIM 607854), the gene mutant in vitelliform macular dystrophy (VMD; MIM 153700), and 2 other BEST1-like genes, BEST2 (MIM 607335) and BEST4 (MIM 607336). Bestrophins are transmembrane (TM) proteins that share a homology region containing a high content of aromatic residues, including an invariant arg-phe-pro (RFP) motif. The bestrophin genes share a conserved gene structure, with almost identical sizes of the 8 RFP-TM domain-encoding exons and highly conserved exon-intron boundaries. Each of the 4 bestrophin genes has a unique 3-prime end of variable length (Stohr et al., 2002 [PubMed 12032738]; Tsunenari et al., 2003 [PubMed 12907679]).[supplied by OMIM]
Sequence: YEKHGHRHSMIRRVKRFLSAHEHPSSPRRRSYRRQTSDSSMFLPRDDLSPARDLLDVPSRNPPRASPTWKKSCFPEGSPTLHFSMGELSTIRETSQTSTSpecifications
NP_689652 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YEKHGHRHSMIRRVKRFLSAHEHPSSPRRRSYRRQTSDSSMFLPRDDLSPARDLLDVPSRNPPRASPTWKKSCFPEGSPTLHFSMGELSTIRETSQTST | |
RUO | |
BEST3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
144453 | |
VMD2L3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC13168/MGC40411/VMD2L3 | |
BEST3 | |
Recombinant | |
wheat germ expression system |