missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human VISTA/B7-H5/PD-1H DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
Pack of 5
Description
The Human VISTA/B7-H5/PD-1H DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human VISTA/B7-H5/PD-1H.
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Binding Target | VISTA/B7-H5/PD-1H |
| Assay Range | 23.40 to 1500 pg/mL |
| Format | 96-well |
| Gene ID (Entrez) | 64115 (Human); 74048 (Mouse); 690899 (Rat); 102116468 (Cynomolgus Monkey) |
| Sample Type | Cell Culture Supernatants, Serum, EDTA Plasma, Heparin Plasma |
| Synonym | 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1 H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction