missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human VEGFB Partial ORF (NP_003368, 27 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007423-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: DAPGHQRKVVSWIDVYTRATCQPREVVVPLTVELMGTVAKQLVPSCVTVQRCGGCCPDDGLECVPTGQHQVRMQILMIRYPSSQLGEMSLEEHSQCECRPKKKDSAVKPDSpecifications
NP_003368 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DAPGHQRKVVSWIDVYTRATCQPREVVVPLTVELMGTVAKQLVPSCVTVQRCGGCCPDDGLECVPTGQHQVRMQILMIRYPSSQLGEMSLEEHSQCECRPKKKDSAVKPD | |
RUO | |
VEGFB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7423 | |
VEGFB (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
VEGFL/VRF | |
VEGFB | |
Recombinant | |
wheat germ expression system |