Learn More
Abnova™ Human UVRAG Partial ORF (NP_003360.2, 601 a.a. - 699 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007405-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene complements the ultraviolet sensitivity of xeroderma pigmentosum group C cells and encodes a protein with a C2 domain. The protein activates the Beclin1-PI(3)KC3 complex, promoting autophagy and suppressing the proliferation and tumorigenicity of human colon cancer cells. Chromosomal aberrations involving this gene are associated with left-right axis malformation and mutations in this gene have been associated with colon cancer. [provided by RefSeq]
Sequence: LPSEQAGSASVQLPGEFHPVSEAELCCTVEQAEEIIGLEATGFASGDQLEAFNCIPVDSAVAVECDEQVLGEFEEFSRRIYALNENVSSFRRPRRSSDKSpecifications
NP_003360.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LPSEQAGSASVQLPGEFHPVSEAELCCTVEQAEEIIGLEATGFASGDQLEAFNCIPVDSAVAVECDEQVLGEFEEFSRRIYALNENVSSFRRPRRSSDK | |
RUO | |
UVRAG | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7405 | |
UVRAG (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DHTX/p63 | |
UVRAG | |
Recombinant | |
wheat germ expression system |