missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UVRAG Partial ORF (NP_003360.2, 601 a.a. - 699 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007405-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107385

  • 508.00€ / 25µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene complements the ultraviolet sensitivity of xeroderma pigmentosum group C cells and encodes a protein with a C2 domain. The protein activates the Beclin1-PI(3)KC3 complex, promoting autophagy and suppressing the proliferation and tumorigenicity of human colon cancer cells. Chromosomal aberrations involving this gene are associated with left-right axis malformation and mutations in this gene have been associated with colon cancer. [provided by RefSeq]

Sequence: LPSEQAGSASVQLPGEFHPVSEAELCCTVEQAEEIIGLEATGFASGDQLEAFNCIPVDSAVAVECDEQVLGEFEEFSRRIYALNENVSSFRRPRRSSDK
Specifications

Specifications

NP_003360.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LPSEQAGSASVQLPGEFHPVSEAELCCTVEQAEEIIGLEATGFASGDQLEAFNCIPVDSAVAVECDEQVLGEFEEFSRRIYALNENVSSFRRPRRSSDK
RUO
UVRAG
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7405
UVRAG (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DHTX/p63
UVRAG
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers