Learn More
Abnova™ Human USP46 Full-length ORF (NP_073743.2, 1 a.a. - 366 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064854-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP46 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes (Quesada et al., 2004 [PubMed 14715245]).[supplied by OMIM]
Sequence: MTVRNIASICNMGTNASALEKDIGPEQFPINEHYFGLVNFGNTCYCNSVLQALYFCRPFRENVLAYKAQQKKKENLLTCLADLFHSIATQKKKVGVIPPKKFISRLRKENDLFDNYMQQDAHEFLNYLLNTIADILQEEKKQEKQNGKLKNGNMNEPAENNKPELTWVHEIFQGTLTNETRCLNCETVSSKDEDFLDLSVDVEQNTSITHCLRDFSNTETLCSEQKYYCETCCSKQEAQKRMRVKKLPMILALHLKRFKYMEQLHRYTKLSYRVVFPLELRLFNTSSDAVNLDRMYDLVAVVVHCGSGPNRGHYITIVKSHGFWLLFDDDIVEKIDAQAIEEFYGLTSDISKNSESGYILFYQSRESpecifications
NP_073743.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11850/FLJ12552/FLJ14283/FLJ39393 | |
USP46 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64854 | |
USP46 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTVRNIASICNMGTNASALEKDIGPEQFPINEHYFGLVNFGNTCYCNSVLQALYFCRPFRENVLAYKAQQKKKENLLTCLADLFHSIATQKKKVGVIPPKKFISRLRKENDLFDNYMQQDAHEFLNYLLNTIADILQEEKKQEKQNGKLKNGNMNEPAENNKPELTWVHEIFQGTLTNETRCLNCETVSSKDEDFLDLSVDVEQNTSITHCLRDFSNTETLCSEQKYYCETCCSKQEAQKRMRVKKLPMILALHLKRFKYMEQLHRYTKLSYRVVFPLELRLFNTSSDAVNLDRMYDLVAVVVHCGSGPNRGHYITIVKSHGFWLLFDDDIVEKIDAQAIEEFYGLTSDISKNSESGYILFYQSRE | |
RUO | |
USP46 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |