missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human USP42 Partial ORF (XP_499256, 601 a.a. - 710 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084132-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: SSVLVPYGAESSEDSDEESKGLGKENGIGTIVSSHSPGQDAEDEEATPHELQEPMTLNGANSADSDSDPKENGLAPDGASCQGQPALHSENPFAKANGLPGKLMPAPLLSSpecifications
XP_499256 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SSVLVPYGAESSEDSDEESKGLGKENGIGTIVSSHSPGQDAEDEEATPHELQEPMTLNGANSADSDSDPKENGLAPDGASCQGQPALHSENPFAKANGLPGKLMPAPLLS | |
RUO | |
USP42 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84132 | |
USP42 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12697 | |
USP42 | |
Recombinant | |
wheat germ expression system |