missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human USP37 Partial ORF (NP_065986, 123 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057695-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102913

  • 528.00€ / 25µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

Sequence: ILGSRTSQKETSRQLSYSDNQASAKRGSLETKDDIPFRKVLGNPGRGSIKTVAGSGIARTIPSLTSTSTPLRSGLLENRTEKRKRMISTGSELNEDYPKENDSSSN
Specifications

Specifications

NP_065986
Liquid
57695
USP37 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1594/MGC117261
USP37
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.4kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ILGSRTSQKETSRQLSYSDNQASAKRGSLETKDDIPFRKVLGNPGRGSIKTVAGSGIARTIPSLTSTSTPLRSGLLENRTEKRKRMISTGSELNEDYPKENDSSSN
RUO
USP37
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers