missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UNC5B Partial ORF (NP_734465, 27 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00219699-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16162447

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

UNC5B belongs to a family of netrin-1 (MIM 601614) receptors thought to mediate the chemorepulsive effect of netrin-1 on specific axons. For more information on UNC5 proteins, see UNC5C (MIM 603610).[supplied by OMIM]

Sequence: GTDSGSEVLPDSFPSAPAEPLPYFLQEPQDAYIVKNKPVELRCRAFPATQIYFKCNGEWVSQNDHVTQEGLDEATGLRVREVQIEVSRQQVEELFGLEDY
Specifications

Specifications

NP_734465
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GTDSGSEVLPDSFPSAPAEPLPYFLQEPQDAYIVKNKPVELRCRAFPATQIYFKCNGEWVSQNDHVTQEGLDEATGLRVREVQIEVSRQQVEELFGLEDY
RUO
UNC5B
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
219699
UNC5B (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
UNC5H2/p53RDL1
UNC5B
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers