Learn More
Abnova™ Human UCHL5IP Full-length ORF (NP_059988.3, 1 a.a. - 368 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055559-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein identified by interaction with ubiquitin C-terminal hydrolase 37, which functions to edit polyubiquitin chains on ubiquitinated substrates. This protein is a subunit of the multisubunit augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. [provided by RefSeq]
Sequence: MGGARLGARNMAGQDAGCGRGGDDYSEDEGDSSVSRAAVEVFGKLKDLNCPFLEGLYITEPKTIQELLCSPSEYRLEILEWMCTRVWPSLQDRFSSLKGVPTEVKIQEMTKLGHELMLCAPDDQELLKGCACAQKQLHFMDQLLDTIRSLTIGCSSCSSLMEHFEDTREKNEALLGELFSSPHLQMLLNPECDPWPLDMQPLLNKQSDDWQWASASAKSEEEEKLAELARQLQESAAKLHALRTEYFAQHEQGAAAGAADISTLDQKLRLVTSDFHQLILAFLQVYDDELGECCQRPGPDLHPCGPIIQATHQNLTSYSQLLQVVMAVADTSAKAVETVKKQQGEQICWGGSSSVMSLATKMNELMEKSpecifications
NP_059988.3 | |
Liquid | |
55559 | |
UCHL5IP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGGARLGARNMAGQDAGCGRGGDDYSEDEGDSSVSRAAVEVFGKLKDLNCPFLEGLYITEPKTIQELLCSPSEYRLEILEWMCTRVWPSLQDRFSSLKGVPTEVKIQEMTKLGHELMLCAPDDQELLKGCACAQKQLHFMDQLLDTIRSLTIGCSSCSSLMEHFEDTREKNEALLGELFSSPHLQMLLNPECDPWPLDMQPLLNKQSDDWQWASASAKSEEEEKLAELARQLQESAAKLHALRTEYFAQHEQGAAAGAADISTLDQKLRLVTSDFHQLILAFLQVYDDELGECCQRPGPDLHPCGPIIQATHQNLTSYSQLLQVVMAVADTSAKAVETVKKQQGEQICWGGSSSVMSLATKMNELMEK | |
RUO | |
UCHL5IP | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSXQ28ORF/UIP1 | |
UCHL5IP | |
Yes | |
wheat germ expression system |