Learn More
Abnova™ Human UBR2 Full-length ORF (AAH64512.1, 1 a.a. - 439 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023304-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Proteolysis by the ubiquitin-proteasome system controls the concentration of many regulatory proteins. The selectivity of ubiquitylation is determined by ubiquitin E3 ligases, which recognize the substrate's destabilization signal, or degron. The E3 ligase UBR2 participates in the N-end rule pathway, which targets proteins bearing an N-terminal degron, or N-degron (Kwon et al., 2003 [PubMed 14585983]).[supplied by OMIM]
Sequence: MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCGRVFKVGEPTYSCRDCAVDPTCVLCMECFLGSIHRDHRYRMTTSGGGGFCDCGDTEAWKEGPYCQKHELNTSEIEEEEDPLVHLSEDVIARTYNIFAITFRYAVEILTWEKESELPADLEMVEKSDTYYCMLFNDEVHTYEQVIYTLQKAVNCTQKEAIGFATTVDRDGRRSVRYGDFQYCEQAKSVIVRNTSRQTKPLKVQVMHSSIVAHQNFGLKLLSWLGSIIGYSDGLRRILCQVGLQEGPDGENSSLVDRLMLSDSKLWKGARSVYHQLFMSSLLMDLKYKKLFAVRFAKNYERLQSDYVTDDHDREFSVADLSVQIFTVPSLFSISAGRSGSPLSpecifications
AAH64512.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf133/DKFZp686C08114/KIAA0349/MGC71112/RP3-392M17.3/bA49A4.1/dJ242G1.1/dJ392M17.3 | |
UBR2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23304 | |
UBR2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASELEPEVQAIDRSLLECSAEEIAGKWLQATDLTREVYQHLAHYVPKIYCRGPNPFPQKEDMLAQHVLLGPMEWYLCGEDPAFGFPKLEQANKPSHLCGRVFKVGEPTYSCRDCAVDPTCVLCMECFLGSIHRDHRYRMTTSGGGGFCDCGDTEAWKEGPYCQKHELNTSEIEEEEDPLVHLSEDVIARTYNIFAITFRYAVEILTWEKESELPADLEMVEKSDTYYCMLFNDEVHTYEQVIYTLQKAVNCTQKEAIGFATTVDRDGRRSVRYGDFQYCEQAKSVIVRNTSRQTKPLKVQVMHSSIVAHQNFGLKLLSWLGSIIGYSDGLRRILCQVGLQEGPDGENSSLVDRLMLSDSKLWKGARSVYHQLFMSSLLMDLKYKKLFAVRFAKNYERLQSDYVTDDHDREFSVADLSVQIFTVPSLFSISAGRSGSPL | |
RUO | |
UBR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |