Learn More
Abnova™ Human UBE2Z Full-length ORF (NP_075567.1, 1 a.a. - 246 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00065264-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Modification of proteins with ubiquitin (UBB; MIM 191339) or ubiquitin-like proteins controls many signaling networks and requires a ubiquitin activating enzyme (E1), a ubiquitin conjugating enzyme (E2), and a ubiquitin protein ligase (E3). UBE2Z is an E2 enzyme (Jin et al., 2007 [PubMed 17597759]).[supplied by OMIM]
Sequence: MSIYKEPPPGMFVVPDTVDMTKIHALITGPFDTPYEGGFFLFVFRCPPDYPIHPPRVKLMTTGNNTVRFNPNFYRNGKVCLSILGTWTGPAWSPAQSISSVLISIQSLMTENPYHNEPGFEQERHPGDSKNYNECIRHETIRVAVCDMMEGKCPCPEPLRGVMEKSFLEYYDFYEVACKDRLHLQGQTMQDPFGEKRGHFDYQSLLMRLGLIRQKVLERLHNENAEMDSDSSSSGTETDLHGSLRVSpecifications
NP_075567.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ13855/HOYS7/USE1 | |
UBE2Z | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
65264 | |
UBE2Z (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSIYKEPPPGMFVVPDTVDMTKIHALITGPFDTPYEGGFFLFVFRCPPDYPIHPPRVKLMTTGNNTVRFNPNFYRNGKVCLSILGTWTGPAWSPAQSISSVLISIQSLMTENPYHNEPGFEQERHPGDSKNYNECIRHETIRVAVCDMMEGKCPCPEPLRGVMEKSFLEYYDFYEVACKDRLHLQGQTMQDPFGEKRGHFDYQSLLMRLGLIRQKVLERLHNENAEMDSDSSSSGTETDLHGSLRV | |
RUO | |
UBE2Z | |
Wheat Germ (in vitro) | |
GST | |
Liquid |