missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UBE1 Partial ORF (NP_003325, 751 a.a. - 850 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_003325 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7317 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16167175
|
Abnova™
H00007317-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16157175
|
Abnova™
H00007317-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11.23. Alternatively spliced transcript variants that encode the same protein have been described. [provided by RefSeq]
Sequence: PLTFDVNNPLHLDYVMAAANLFAQTYGLTGSQDRAAVATFLQSVQVPEFTPKSGVKIHVSDQELQSANASVDDSRLEELKATLPSPDKLPGFKMYPIDFESpecifications
NP_003325 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
A1S9/A1S9T/A1ST/AMCX1/GXP1/MGC4781/SMAX2/UBA1A/UBE1/UBE1X | |
UBA1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7317 | |
UBE1 (Human) Recombinant Protein (Q01) | |
PLTFDVNNPLHLDYVMAAANLFAQTYGLTGSQDRAAVATFLQSVQVPEFTPKSGVKIHVSDQELQSANASVDDSRLEELKATLPSPDKLPGFKMYPIDFE | |
RUO | |
UBA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |