missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UBASH3A Full-length ORF (AAH28138.1, 1 a.a. - 451 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00053347-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120393

  • 335.00€ / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MAAGETQLYAKVSNKLKGRSSPSLLEPLLAMGFPVHTALKALAATGRKTAEEALAWLHDHCNDPSLDDPIPQEYALFLCPTGPLLEKLQEFWRESKRQCAKNRAHEVFPHVTLCDFFTCEDQKVECLYEALKRAGDRLLGSFPTAVPLALHSSISYLGFFVSGSPADVIREFAMTFATEASLLADCSVKPCTKQLHLTLAHKFYPHHQRTLEQLARAIPLGHSCQWTAALYSRDMRFVHYQTLRALFQYKPQNVDELTLSPGDYIFVDPTQQDEASEGWVIGISQRTGCRGFLPENYTDRASESDTWVKHRMYTFSLATDLNSRKDGEASSRCSGEFLPQTARSLSSLQALQATVARKSVLVVRHGERVDQIFGKAWLQQCSTPDGNYYRPDLNFPCSLPRRSRGIKDFENDPPLSSCGIFQSRIAGMFEDCLVGKVTITATLIMASRHPA
Specifications

Specifications

AAH28138.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLIP4/STS-2/TULA
UBASH3A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
53347
UBASH3A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAGETQLYAKVSNKLKGRSSPSLLEPLLAMGFPVHTALKALAATGRKTAEEALAWLHDHCNDPSLDDPIPQEYALFLCPTGPLLEKLQEFWRESKRQCAKNRAHEVFPHVTLCDFFTCEDQKVECLYEALKRAGDRLLGSFPTAVPLALHSSISYLGFFVSGSPADVIREFAMTFATEASLLADCSVKPCTKQLHLTLAHKFYPHHQRTLEQLARAIPLGHSCQWTAALYSRDMRFVHYQTLRALFQYKPQNVDELTLSPGDYIFVDPTQQDEASEGWVIGISQRTGCRGFLPENYTDRASESDTWVKHRMYTFSLATDLNSRKDGEASSRCSGEFLPQTARSLSSLQALQATVARKSVLVVRHGERVDQIFGKAWLQQCSTPDGNYYRPDLNFPCSLPRRSRGIKDFENDPPLSSCGIFQSRIAGMFEDCLVGKVTITATLIMASRHPA
RUO
UBASH3A
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers