Learn More
Abnova™ Human UBA7 Full-length ORF (NP_003326.2, 1 a.a. - 1012 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_003326.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7318 |
Molecular Weight (g/mol) | 138.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16161672
|
Abnova™
H00007318-P02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16151672
|
Abnova™
H00007318-P02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E1 ubiquitin-activating enzyme family. The encoded enzyme is a retinoid target that triggers promyelocytic leukemia (PML)/retinoic acid receptor alpha (RARalpha) degradation and apoptosis in acute promyelocytic leukemia, where it is involved in the conjugation of the ubiquitin-like interferon-stimulated gene 15 protein. [provided by RefSeq]
Sequence: MDALDASKLLDEELYSRQLYVLGSPAMQRIQGARVLVSGLQGLGAEVAKNLVLMGVGSLTLHDPHPTCWSDLAAQFLLSEQDLERSRAEASQELLAQLNRAVQVVVHTGDITEDLLLDFQVVVLTAAKLEEQLKVGTLCHKHGVCFLAADTRGLVGQLFCDFGEDFTVQDPTEAEPLTAAIQHISQGSPGILTLRKGANTHYFRDGDLVTFSGIEGMVELNDCDPRSIHVREDGSLEIGDTTTFSRYLRGGAITEVKRPKTVRHKSLDTALLQPHVVAQSSQEVHHAHCLHQAFCALHKFQHLHGRPPQPWDPVDAETVVGLARDLEPLKRTEEEPLEEPLDEALVRTVALSSAGVLSPMVAMLGAVAAQEVLKAISRKFMPLDQWLYFDALDCLPEDGELLPSPEDCALRGSRYDGQIAVFGAGFQEKLRRQHYLLVGAGAIGCELLKVFALVGLGAGNSGGLTVVDMDHIERSNLSRQFLFRSQDVGRPKAEVAAAAARGLNPDLQVIPLTYPLDPTTEHIYGDNFFSRVDGVAAALDSFQARRYVAARCTHYLKPLLEAGTSGTWGSATVFMPHVTEAYRAPASAAASEDAPYPVCTVRYFPSTAEHTLQWARHEFEELFRLSAETINHHQQAHTSLADMDEPQTLTLLKPVLGVLRVRPQNWQDCVAWALGHWKLCFHYGIKQLLRHFPPNKVLEDGTPFWSGPKQCPQPLEFDTNQDTHLLYVLAAANLYAQMHGLPGSQDWTALRELLKLLPQPDPQQMAPIFASNLELASASAEFGPEQQKELNKALEVWSVGPPLKPLMFEKDDDSNFHVDFVVAAASLRCQNYGIPPVNRAQSKRIVGQIIPAIATTTAAVAGLLGLELYKVVSGPRPRSAFRHSYLHLAENYLIRYMPFAPAIQTFHHLKWTSWDRLKVPAGQPERTLESLLAHLQEQHGLRVRILLHGSALLYAAGWSPEKQAQHLPLRVTELVQQLTGQAPAPGQRVLVLELSCEGDDEDTAFPPLHYELSpecifications
NP_003326.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
138.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDALDASKLLDEELYSRQLYVLGSPAMQRIQGARVLVSGLQGLGAEVAKNLVLMGVGSLTLHDPHPTCWSDLAAQFLLSEQDLERSRAEASQELLAQLNRAVQVVVHTGDITEDLLLDFQVVVLTAAKLEEQLKVGTLCHKHGVCFLAADTRGLVGQLFCDFGEDFTVQDPTEAEPLTAAIQHISQGSPGILTLRKGANTHYFRDGDLVTFSGIEGMVELNDCDPRSIHVREDGSLEIGDTTTFSRYLRGGAITEVKRPKTVRHKSLDTALLQPHVVAQSSQEVHHAHCLHQAFCALHKFQHLHGRPPQPWDPVDAETVVGLARDLEPLKRTEEEPLEEPLDEALVRTVALSSAGVLSPMVAMLGAVAAQEVLKAISRKFMPLDQWLYFDALDCLPEDGELLPSPEDCALRGSRYDGQIAVFGAGFQEKLRRQHYLLVGAGAIGCELLKVFALVGLGAGNSGGLTVVDMDHIERSNLSRQFLFRSQDVGRPKAEVAAAAARGLNPDLQVIPLTYPLDPTTEHIYGDNFFSRVDGVAAALDSFQARRYVAARCTHYLKPLLEAGTSGTWGSATVFMPHVTEAYRAPASAAASEDAPYPVCTVRYFPSTAEHTLQWARHEFEELFRLSAETINHHQQAHTSLADMDEPQTLTLLKPVLGVLRVRPQNWQDCVAWALGHWKLCFHYGIKQLLRHFPPNKVLEDGTPFWSGPKQCPQPLEFDTNQDTHLLYVLAAANLYAQMHGLPGSQDWTALRELLKLLPQPDPQQMAPIFASNLELASASAEFGPEQQKELNKALEVWSVGPPLKPLMFEKDDDSNFHVDFVVAAASLRCQNYGIPPVNRAQSKRIVGQIIPAIATTTAAVAGLLGLELYKVVSGPRPRSAFRHSYLHLAENYLIRYMPFAPAIQTFHHLKWTSWDRLKVPAGQPERTLESLLAHLQEQHGLRVRILLHGSALLYAAGWSPEKQAQHLPLRVTELVQQLTGQAPAPGQRVLVLELSCEGDD | |
RUO | |
UBA7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7318 | |
UBA7 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D8/MGC12713/UBA1B/UBE1L/UBE2 | |
UBA7 | |
Recombinant | |
wheat germ expression system |