missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human U2AF1L3 Full-length ORF (AAH21186.1, 1 a.a. - 202 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00199746-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MAEYLASIFGTEKDKVNCSFYFKIGVCRHGDRCSRLHNKPTFSQEVFTELQEKYGEIEEMNVCDNLGDHVVGNVYVKFRREEDGERAVAELSNRWFNGQAVHGNVPEVASATSCICGPFPRTSRGSSMGGDPGAGHPRGSILATIPERGTIGVPLITGMAASEALAPLPFTPNRDRCSWQDLSSKPPSLSCPILPRLPGSIMSpecifications
AAH21186.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ35525/MGC33901/U2AF1-RS3/U2AF1L3/U2AF1L3V1/U2AF1RS3/U2af26 | |
U2AF1L4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
199746 | |
U2AF1L3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEYLASIFGTEKDKVNCSFYFKIGVCRHGDRCSRLHNKPTFSQEVFTELQEKYGEIEEMNVCDNLGDHVVGNVYVKFRREEDGERAVAELSNRWFNGQAVHGNVPEVASATSCICGPFPRTSRGSSMGGDPGAGHPRGSILATIPERGTIGVPLITGMAASEALAPLPFTPNRDRCSWQDLSSKPPSLSCPILPRLPGSIM | |
RUO | |
U2AF1L4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |