missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TUSC3 Full-length ORF (NP_839952.1, 1 a.a. - 347 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007991-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192232

  • 528.00€ / 25µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

This gene is a candidate tumor suppressor gene. It is located within a homozygously deleted region of a metastatic prostate cancer. The gene is expressed in most nonlymphoid human tissues including prostate, lung, liver, and colon. Expression was also detected in many epithelial tumor cell lines. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MGARGAPSRRRQAGRRLRYLPTGSFPFLLLLLLLCIQLGGGQKKKENLLAEKVEQLMEWSSRRSIFRMNGDKFRKFIKAPPRNYSMIVMFTALQPQRQCSVCRQANEEYQILANSWRYSSAFCNKLFFSMVDYDEGTDVFQQLNMNSAPTFMHFPPKGRPKRADTFDLQRIGFAAEQLAKWIADRTDVHIRVFRPPNYSGTIALALLVSLVGGLLYLRRNNLEFIYNKTGWAMVSLCIVFAMTSGQMWNHIRGPPYAHKNPHNGQVSYIHGSSQAQFVAESHIILVLNAAITMGMVLLNEAATSKGDVGKRRIICLVGLGLVVFFFSFLLSIFRSKYHGYPYSFLIK
Specifications

Specifications

NP_839952.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
66kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
D8S1992/M33/MGC13453/MRT7/N33/OST3A
TUSC3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7991
TUSC3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGARGAPSRRRQAGRRLRYLPTGSFPFLLLLLLLCIQLGGGQKKKENLLAEKVEQLMEWSSRRSIFRMNGDKFRKFIKAPPRNYSMIVMFTALQPQRQCSVCRQANEEYQILANSWRYSSAFCNKLFFSMVDYDEGTDVFQQLNMNSAPTFMHFPPKGRPKRADTFDLQRIGFAAEQLAKWIADRTDVHIRVFRPPNYSGTIALALLVSLVGGLLYLRRNNLEFIYNKTGWAMVSLCIVFAMTSGQMWNHIRGPPYAHKNPHNGQVSYIHGSSQAQFVAESHIILVLNAAITMGMVLLNEAATSKGDVGKRRIICLVGLGLVVFFFSFLLSIFRSKYHGYPYSFLIK
RUO
TUSC3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers