Learn More
R&D Systems™ Human TSLP DuoSet ELISA
Product Code. 30021258
Click to view available options
Format:
15 96-well Plates
5 96-well Plates
Unit Size:
1 set
Pack of 5
Product Code. 30021258
Brand: R&D Systems™ DY1398
Human TSLP DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human TSLP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TSLP.
TRUSTED_SUSTAINABILITY
Specifications
Product Type ELISA Kit
Quantity 1 Kit
Species Human
Conjugate Biotin
Assay Range 31.20 to 2000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 15 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 85480
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.